Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl glutarate, 98%, Thermo Scientific™

Catalog No. AAL1125914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1125914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1125914 Supplier Thermo Scientific Chemicals Supplier No. L1125914

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-38-2
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00009212
InChI Key OUWSNHWQZPEFEX-UHFFFAOYSA-N
Synonym diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters
PubChem CID 13163
ChEBI CHEBI:87319
IUPAC Name diethyl pentanedioate
SMILES CCOC(=O)CCCC(=O)OCC

Specifications

Density 1.022
Boiling Point 236°C to 237°C
Flash Point 96°C (204°F)
Refractive Index 1.423
Quantity 25 g
Beilstein 1777327
Formula Weight 188.23
Percent Purity 98%
Chemical Name or Material Diethyl glutarate
EINECSNumber 212-451-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.