Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Glutarate 98.0+%, TCI America™
SDP

Catalog No. G0184500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0184500G 500 g
G018425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0184500G Supplier TCI America Supplier No. G0184500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 818-38-2
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00009212
InChI Key OUWSNHWQZPEFEX-UHFFFAOYSA-N
Synonym diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters
PubChem CID 13163
ChEBI CHEBI:87319
IUPAC Name 1,5-diethyl pentanedioate
SMILES CCOC(=O)CCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 230°C
Quantity 500 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Glutarate
EINECSNumber (2)-3088
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.