Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (Hydroxymethyl)Phosphonate

Catalog No. 39262625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-262-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-262-625ML Supplier Sigma Aldrich Supplier No. 39262625ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of:+a And +a-dialkoxyphosphoryl isothiocyanate used in antIpr∗

Specifications

CAS 17049-50-2
Density 0.846 g/mL at 25°C
Molecular Formula C10H21BrMg
Refractive Index n20/D 1.444
Linear Formula CH3(CH2)9MgBr
MDL Number MFCD00013516
Quantity 25 mL
Synonym (Hydroxymethyl)phosphonic acid diethyl ester; Diethyl phosphonomethanol
Molecular Weight (g/mol) 245.48 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.