Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl iminodiacetate, 97%

Catalog No. AAH60608AC
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60608AC 25 mL
AAH60608AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60608AC Supplier Thermo Scientific Chemicals Supplier No. H60608AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol

It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-05-7
MDL Number MFCD00041925

Specifications

Density 1.056
Quantity 25 mL
Flash Point 113°C (235°F)
Refractive Index 1.435
Molecular Formula C8H15NO4
Linear Formula HN(CH2CO2C2H5)2
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
Boiling Point 208°C
Solubility Information Moderately soluble in water.
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
SMILES CCOC(=O)CNCC(=O)OCC
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol) 189.21
PubChem CID 80502
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material Diethyl iminodiacetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-533-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.