Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Iminodiacetate 98.0+%, TCI America™
SDP

Catalog No. I032625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I032625ML 25 mL
I03265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I032625ML Supplier TCI America Supplier No. I032625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6290-05-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00041925
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
PubChem CID 80502
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
SMILES CCOC(=O)CNCC(=O)OCC

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 mL
Formula Weight 189.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Iminodiacetate
EINECSNumber (2)-3117
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.