Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Isopropylidenesuccinate 98.0+%, TCI America™
SDP

Catalog No. I04041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04041G 1 g
I04045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04041G Supplier TCI America Supplier No. I04041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42103-98-0
Molecular Formula C11H18O4
Molecular Weight (g/mol) 214.261
MDL Number MFCD00130132
InChI Key FSUBDXPWXDGTBG-UHFFFAOYSA-N
Synonym Isopropylidenesuccinic Acid Diethyl Ester
PubChem CID 251287
IUPAC Name diethyl 2-propan-2-ylidenebutanedioate
SMILES CCOC(=O)CC(=C(C)C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 103°C
Quantity 1 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Isopropylidenesuccinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.