Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Isopropylmalonate 98.0+%, TCI America™
SDP

Catalog No. I025225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025225ML 25 mL
I0252100ML 100 mL
I0252500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I025225ML Supplier TCI America Supplier No. I025225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 759-36-4
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00040491
InChI Key BYQFBFWERHXONI-UHFFFAOYSA-N
Synonym diethyl isopropylmalonate, diethyl 2-isopropylmalonate, isopropyl diethyl malonate, 1,3-diethyl 2-propan-2-yl propanedioate, diethylisopropylmalonate, isopropylmalonic acid diethyl ester, propanedioic acid, 1-methylethyl-, diethyl ester, ethyl isopropylmalonate, 1,3-diethyl 2-isopropylpropanedioate, 2-isopropyl-malonic acid diethyl ester
PubChem CID 12966
IUPAC Name 1,3-diethyl 2-(propan-2-yl)propanedioate
SMILES CCOC(=O)C(C(C)C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Isopropylmalonate
EINECSNumber (2)-0879
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.