Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Itaconate (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. I0204500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0204500G 500 g
I020425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0204500G Supplier TCI America Supplier No. I0204500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2409-52-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD00059230
InChI Key ZEFVHSWKYCYFFL-UHFFFAOYSA-N
Synonym Itaconic Acid Diethyl Ester
PubChem CID 75481
IUPAC Name diethyl 2-methylidenebutanedioate
SMILES CCOC(=O)CC(=C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 228°C
Quantity 500 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Itaconate (stabilized with TBC)
EINECSNumber (2)-1130&(2)-1142
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.