Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ketomalonate, 95%

Catalog No. AC114430050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114430050 5 g Glass bottle
AC114430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114430050 Supplier Thermo Scientific Chemicals Supplier No. 114430050
Add to Cart

CAS: 609-09-6 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-09-6
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00009121
InChI Key DBKKFIIYQGGHJO-UHFFFAOYSA-N
Synonym diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester
PubChem CID 69105
IUPAC Name diethyl 2-oxopropanedioate
SMILES CCOC(=O)C(=O)C(=O)OCC

Specifications

Color Yellow
Density 1.1420g/mL
Boiling Point 208.0°C to 210.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4150 to 1.4190
Linear Formula C2H5O2CCOCO2C2H5
Quantity 5 g
Beilstein 03, 769
Fieser 06,188; 08,170
Specific Gravity 1.142
Merck Index 15, 5983
Formula Weight 174.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl ketomalonate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 210-176-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.