Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Diethyl L-Tartrate

Catalog No. 156841100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-684-1100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-684-1100G Supplier Sigma Aldrich Supplier No. 156841100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Made from natural tartaric acid(+)-Diethyl L-tartrate can be used in the synthesis of biologically active compounds such as (+)-altholactone, (-)-aspicilin, (+)-monomorine I and (+)-(1R,2R,3S,6S)-3,6-di-O-methyl conduritol-E.

Specifications

CAS 87-91-2
Melting Point 150°C to 152°C
Molecular Formula C18H22N2O6
Refractive Index n20/D 1.446
Linear Formula C18H22N2O6
MDL Number MFCD00037911
Quantity 5 g
Synonym L-(+)-Tartaric acid diethyl ester
Molecular Weight (g/mol) 362.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.