Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl malonate

Catalog No. D9775425KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9775425KG 2.5 kg
D9775425G 25 g
D977541KG 1 kg
D97754500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D9775425KG Supplier Sigma Aldrich Supplier No. D977542.5KG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl malonate

Specifications

CAS 105-53-3
Melting Point -51°C to +50°C (lit.)
Density 1.055 g/mL (at 25°C (literature))
Boiling Point 199°C (lit.)
Molecular Formula C7H12O4
Refractive Index n20/D 1.413 (literature)
Linear Formula CH2(COOC2H5)2
MDL Number MFCD00009195
Quantity 2.5 kg
Synonym Malonic acid diethyl ester
Molecular Weight (g/mol) 160.17
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.