Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Methylphosphonate

Catalog No. 2681195G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-811-95G 5 g
26-811-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 26-811-95G Supplier Sigma Aldrich Supplier No. 2681195G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent employed in the synthesis of +a-alkenyl, +a-hydroxyl, and dialkyl fluoroalkynyl phosphonates.

Specifications

CAS 683-08-9
Density 1.041 g/mL at 25°C
Boiling Point 194°C
Molecular Formula C5H13O3P
Refractive Index n20/D 1.414
Linear Formula CH3P(O)(OC2H5)2
MDL Number MFCD00009813
Quantity 5 g
Synonym Bis(diethoxyphosphinyl)methane; DEMP; Diethoxymethylphosphine oxide
Molecular Weight (g/mol) 152.13 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.