Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Oxalate, 99%, Reagents

Catalog No. 01337285
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-285 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-285 Supplier Reagents Holdings Llc Supplier No. C2195960500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl Oxalate, 99%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 95-92-1
Boiling Range 185°C
Melting Point -41°C
Color Colorless
Boiling Point 185°C
Melting Range -41°C
Packaging Glass Bottle
Molecular Formula C6H10O4
Quantity 500 mL
Specific Gravity 1.079
Synonym Ethyl Oxalate, Diethyl Ethanedioate, Ethanedioic Acid, Diethyl Ester
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
SMILES CCOC(=O)C(=O)OCC
IUPAC Name diethyl oxalate
Molecular Weight (g/mol) 146.14
Grade Reagent
Physical Form Liquid
Chemical Name or Material Diethyl Oxalate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.