Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Oxalate 99.0+%, TCI America™
SDP

Catalog No. O0120250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0120250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0120250ML Supplier TCI America Supplier No. O0120250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95-92-1
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00009119
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
Synonym ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove
PubChem CID 7268
IUPAC Name diethyl oxalate
SMILES CCOC(=O)C(=O)OCC

Specifications

Melting Point -41°C
Color Colorless
Boiling Point 185°C
Quantity 250 mL
UN Number 2525
Formula Weight 146.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Oxalate [for Spectrophotometry]
EINECSNumber (2)-0924
RTECSNumber RO2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.