Learn More
Medchemexpress LLC Diethyl oxalpropionate | 759-65-9 | 95.0% | 1 ML
SDP
Catalog No. 5000340745
Encompass_Preferred
Diethyl oxalpropionate is an intermediate used in the synthesis of poly((R,S)-3,3-dimethylmalic acid) (PDMMLA) derivatives. These PDMMLA derivatives can be utilized in the creation of nanoparticles and for studying warfarin encapsulation and controlled release.
  • Intermediate for PDMMLA derivative synthesis
  • Can be used in the synthesis of nanoparticles
  • Used in the study of warfarin encapsulation and controlled release
Catalog No. 50-003-40745 Supplier Medchemexpress LLC Supplier No. HY12872010MM/1ML
Diethyl oxalpropionate is an intermediate used in the synthesis of poly((R,S)-3,3-dimethylmalic acid) (PDMMLA) derivatives. These PDMMLA derivatives can be utilized in the creation of nanoparticles and for studying warfarin encapsulation and controlled release.
  • Intermediate for PDMMLA derivative synthesis
  • Can be used in the synthesis of nanoparticles
  • Used in the study of warfarin encapsulation and controlled release