Learn More
Medchemexpress LLC Diethyl oxalpropionate | 759-65-9 | 95.0% | 5 G
SDP
Catalog No. 5000340744
Encompass_Preferred
Diethyl oxalpropionate is an intermediate for poly((R,S)-3,3-dimethylmalic acid) (PDMMLA) derivative synthesis. These PDMMLA derivatives can be utilized in the synthesis of nanoparticles and for studying warfarin encapsulation and controlled release.
  • Intermediate for PDMMLA derivative synthesis
  • Utilized in the synthesis of nanoparticles
  • Used for studying warfarin encapsulation
  • Used for studying controlled release
Catalog No. 50-003-40744 Supplier Medchemexpress LLC Supplier No. HY1287205G
Diethyl oxalpropionate is an intermediate for poly((R,S)-3,3-dimethylmalic acid) (PDMMLA) derivative synthesis. These PDMMLA derivatives can be utilized in the synthesis of nanoparticles and for studying warfarin encapsulation and controlled release.
  • Intermediate for PDMMLA derivative synthesis
  • Utilized in the synthesis of nanoparticles
  • Used for studying warfarin encapsulation
  • Used for studying controlled release