Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phenylmalonate, 98%

Catalog No. AC150261000
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150261000 100 mL Glass bottle
AC150262500 250 mL Glass bottle
AC150260010 1 L Glass bottle
AC150260025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150261000 Supplier Thermo Scientific Chemicals Supplier No. 150261000
Add to Cart

CAS: 83-13-6 | C13H16O4 | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-13-6
MDL Number MFCD00009144
InChI Key FGYDHYCFHBSNPE-UHFFFAOYSA-N
Synonym diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
PubChem CID 66514
IUPAC Name diethyl 2-phenylpropanedioate
SMILES CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 16.0°C to 17.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 170.0°C to 172.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C13H16O4
Refractive Index 1.4903 to 1.4923
Linear Formula C2H5O2CCH(C6H5)CO2C2H5
Quantity 100 mL
Beilstein 09, 854
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Diethyl phenylmalonate
Show More Show Less
EINECSNumber 201-456-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.