Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phenylthiomethylphosphonate, Thermo Scientific™

Catalog No. AA1036906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1036906 5 g
AA1036914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1036906 Supplier Thermo Scientific Chemicals Supplier No. 1036906

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38066-16-9
Molecular Formula C11H17O3PS
Molecular Weight (g/mol) 260.288
MDL Number MFCD00015130
InChI Key FBUXEPJJFVDUFE-UHFFFAOYSA-N
Synonym diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate #
PubChem CID 99227
IUPAC Name diethoxyphosphorylmethylsulfanylbenzene
SMILES CCOP(=O)(CSC1=CC=CC=C1)OCC

Specifications

Density 1.174g/mL
Boiling Point 147°C (0.5 mmHg)
Quantity 5 g
Formula Weight 260.3
Physical Form Liquid
Chemical Name or Material Diethyl phenylthiomethylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.