Learn More
Medchemexpress LLC Diethyl phosphate-13C4 sodium | 1329613-90-2 | >95.0% | 180.05 | 13C4H10NaO4P | 1mg
SDP
Catalog No. 5000268495
Encompass_Preferred
Diethyl phosphate-13C4 (sodium) is the 13C labeled isotope of Diethyl phosphate-13C4 (sodium)(HY-101417) Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos[1]
Catalog No. 50-002-68495 Supplier Medchemexpress LLC Supplier No. HYW7781401MG
Diethyl phosphate-13C4 (sodium) is the 13C labeled isotope of Diethyl phosphate-13C4 (sodium)(HY-101417) Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos[1]