Learn More
Apexbio Technology LLC Diethyl phosphate 598-02-7 100mg
SDP
Catalog No. 5000094568
Encompass_Preferred
Diethyl phosphate (CAS 598-02-7) is an organophosphorus compound commonly detected as a metabolic or degradation product of various organophosphate insecticides such as chlorpyrifos While diethyl phosphate itself exhibits low acute toxicity it serves as an important biomarker for organophosphate exposure in environmental and biological monitoring studies The compound does not act via acetylcholinesterase inhibition unlike its parent compounds instead its presence reflects the extent of exposure and metabolic processing of organophosphate pesticides Diethyl phosphate is widely utilized in toxicological environmental and epidemiological research to assess pesticide exposure levels
Catalog No. 50-000-94568 Supplier Apexbio Technology LLC Supplier No. A9972100
Diethyl phosphate (CAS 598-02-7) is an organophosphorus compound commonly detected as a metabolic or degradation product of various organophosphate insecticides such as chlorpyrifos While diethyl phosphate itself exhibits low acute toxicity it serves as an important biomarker for organophosphate exposure in environmental and biological monitoring studies The compound does not act via acetylcholinesterase inhibition unlike its parent compounds instead its presence reflects the extent of exposure and metabolic processing of organophosphate pesticides Diethyl phosphate is widely utilized in toxicological environmental and epidemiological research to assess pesticide exposure levels