Learn More
Medchemexpress LLC Diethyl phosphate | 598-02-7 | MFCD00039889 | 98.1% | 154.10 g/mol | C4H11O4P | 10 G
SDP
Catalog No. 5000245632
Encompass_Preferred
Diethyl phosphate is an organophosphorus reagent (diethyl phosphoric acid) used as a laboratory reagent and analytical standard. It is used in metabolite analysis, chemical synthesis, and as a reference standard for analytical methods. Consult the safety data sheet for hazard classifications, protective measures, and recommended storage conditions.
Catalog No. 50-002-45632 Supplier Medchemexpress LLC Supplier No. HY10141710G
Diethyl phosphate is an organophosphorus reagent (diethyl phosphoric acid) used as a laboratory reagent and analytical standard. It is used in metabolite analysis, chemical synthesis, and as a reference standard for analytical methods. Consult the safety data sheet for hazard classifications, protective measures, and recommended storage conditions.