Learn More
Medchemexpress LLC Diethyl phosphate | 598-02-7 | MFCD00039889 | 98.1% | 154.10 g/mol | C4H11O4P | 500 MG
SDP
Catalog No. 5000249130
Encompass_Preferred
Diethyl phosphate is an organophosphorus compound supplied as an analytical standard and reference material for research applications, including toxicology and environmental metabolite studies. It is commonly used in method development and quantitative/qualitative analyses by HPLC, GC, and MS.
  • Analytical standard suitable for HPLC, GC, and MS.
  • Used as a reference in metabolite and toxicology studies.
  • High purity for reliable quantitative analysis.
  • Available in laboratory-scale pack sizes for convenience.
  • Storage per certificate of analysis preserves stability.
Catalog No. 50-002-49130 Supplier Medchemexpress LLC Supplier No. HY101417500MG
Diethyl phosphate is an organophosphorus compound supplied as an analytical standard and reference material for research applications, including toxicology and environmental metabolite studies. It is commonly used in method development and quantitative/qualitative analyses by HPLC, GC, and MS.
  • Analytical standard suitable for HPLC, GC, and MS.
  • Used as a reference in metabolite and toxicology studies.
  • High purity for reliable quantitative analysis.
  • Available in laboratory-scale pack sizes for convenience.
  • Storage per certificate of analysis preserves stability.