Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyl phosphate-d10-1 | 2483831-48-5 | 99.7% | 164.16 | C4HD10O4P | 5 MG
SDP

Catalog No. 5000247033
Encompass_Preferred

Diethyl phosphate-d10-1 is the deuterium-labeled isotopologue of diethyl phosphate, intended as an analytical reference standard for research and quantitative mass spectrometry. It is supplied as a high-purity liquid suitable for trace-level analyses and isotopic labelling studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity: 99.70%.
  • Molecular weight 164.16 g/mol.
  • Formula C4HD10O4P, suitable for isotopic tracing applications.
  • Colorless to light yellow liquid with defined stability and storage conditions.

Catalog No. 50-002-47033 Supplier Medchemexpress LLC Supplier No. HY101417S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl phosphate-d10-1 is the deuterium-labeled isotopologue of diethyl phosphate, intended as an analytical reference standard for research and quantitative mass spectrometry. It is supplied as a high-purity liquid suitable for trace-level analyses and isotopic labelling studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity: 99.70%.
  • Molecular weight 164.16 g/mol.
  • Formula C4HD10O4P, suitable for isotopic tracing applications.
  • Colorless to light yellow liquid with defined stability and storage conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.