Learn More
Medchemexpress LLC Diethyl phosphoric acid-d10 | 2483831-48-5 | 99.7% | 164.16 g/mol | C4HD10O4P | 1 MG
SDP
Catalog No. 5000249141
Encompass_Preferred
Diethyl phosphate-d10-1 is the deuterium-labeled isotopologue of diethyl phosphoric acid used as an internal standard and reference material for analytical chemistry, particularly mass spectrometry and metabolite quantitation. It is supplied at high purity and is intended for research and analytical applications where isotopic labeling improves assay accuracy.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.7%) for reliable analytical performance.
  • Suitable for quantitative analysis and method development.
  • Available in small sample sizes for analytical workflows.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-49141 Supplier Medchemexpress LLC Supplier No. HY101417S11MG
Diethyl phosphate-d10-1 is the deuterium-labeled isotopologue of diethyl phosphoric acid used as an internal standard and reference material for analytical chemistry, particularly mass spectrometry and metabolite quantitation. It is supplied at high purity and is intended for research and analytical applications where isotopic labeling improves assay accuracy.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.7%) for reliable analytical performance.
  • Suitable for quantitative analysis and method development.
  • Available in small sample sizes for analytical workflows.
  • Certificate of analysis and safety data sheet available.