Learn More
Medchemexpress LLC Diethyl phthalate | 84-66-2 | MFCD00009111 | 99.9% | 222.24 | C12H14O4 | 1kg
SDP
Catalog No. 5000265897
Encompass_Preferred
Diethyl phthalate is an endocrine disruptor that has the activity of affecting the apoptosis system of PC12 cells Diethyl phthalate is widely used in a variety of plastics and personal care products Diethyl phthalate has shown the potential to induce male reproductive toxicity unrelated to androgens in animal experiments[1]
Catalog No. 50-002-65897 Supplier Medchemexpress LLC Supplier No. HYY02841KG
Diethyl phthalate is an endocrine disruptor that has the activity of affecting the apoptosis system of PC12 cells Diethyl phthalate is widely used in a variety of plastics and personal care products Diethyl phthalate has shown the potential to induce male reproductive toxicity unrelated to androgens in animal experiments[1]