Learn More
Medchemexpress LLC Diethyl phthalate | 84-66-2 | MFCD00009111 | 99.9% | 222.24 g/mol | C12H14O4 | 500g
SDP
Catalog No. 5000269981
Encompass_Preferred
Diethyl phthalate is an endocrine disruptor that has the activity of affecting the apoptosis system of PC12 cells Diethyl phthalate is widely used in a variety of plastics and personal care products Diethyl phthalate has shown the potential to induce male reproductive toxicity unrelated to androgens in animal experiments[1]
Catalog No. 50-002-69981 Supplier Medchemexpress LLC Supplier No. HYY0284500G
Diethyl phthalate is an endocrine disruptor that has the activity of affecting the apoptosis system of PC12 cells Diethyl phthalate is widely used in a variety of plastics and personal care products Diethyl phthalate has shown the potential to induce male reproductive toxicity unrelated to androgens in animal experiments[1]