Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Pimelate 99%, Thermo Scientific™

Catalog No. AC114531000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC114531000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC114531000 Supplier Thermo Scientific Chemicals Supplier No. 114531000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-20-6
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00009216
InChI Key LKKOGZVQGQUVHF-UHFFFAOYSA-N
Synonym diethyl pimelate, heptanedioic acid, diethyl ester, diethylpimelate, pimelic acid diethyl ester, unii-2rf63n709k, heptanedioic acid diethyl ester, heptanedioic acid, 1,7-diethyl ester, 1,5-pentanedicarboxylic acid diethyl ester, zlchem 330
PubChem CID 16300
IUPAC Name diethyl heptanedioate
SMILES CCOC(=O)CCCCCC(=O)OCC

Specifications

Color Colorless
Density 0.99g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Linear Formula C2H5O2C(CH2)5CO2C2H5
Quantity 100 g
Beilstein 02, 671
Merck Index 15, 7543
Formula Weight 216.28
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Diethyl pimelate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.