Learn More
Medchemexpress LLC Diethyl propylmalonate | 2163-48-6 | MFCD00009168 | 99.0% | 202.25 g/mol | C10H18O4 | 100 G
SDP
Catalog No. 5000302369
Encompass_Preferred
Diethyl propylmalonate (CAS 2163-48-6) is an organic diester used as a versatile building block in organic synthesis. It is commonly employed in pharmaceutical and agrochemical intermediate synthesis, enabling alkylation and condensation reactions to produce substituted malonates and related derivatives. It is intended for research and small-scale synthetic applications.
Catalog No. 50-003-02369 Supplier Medchemexpress LLC Supplier No. HYW014789100G
Diethyl propylmalonate (CAS 2163-48-6) is an organic diester used as a versatile building block in organic synthesis. It is commonly employed in pharmaceutical and agrochemical intermediate synthesis, enabling alkylation and condensation reactions to produce substituted malonates and related derivatives. It is intended for research and small-scale synthetic applications.