Learn More
Medchemexpress LLC Diethyl propylmalonate | 2163-48-6 | MFCD00009168 | 99.7% | 202.25 g/mol | C10H18O4 | 500 G
SDP
Catalog No. 5000302123
Encompass_Preferred
Diethyl propylmalonate (CAS 2163-48-6) is an organic diester used as a synthetic intermediate in medicinal chemistry and in pesticide synthesis. It is supplied as a high-purity reagent for research, development, and organic synthesis workflows.
  • High purity (99.7%) suitable for synthetic applications.
  • Molecular formula C10H18O4 and molecular weight 202.25 g/mol.
  • Offered in multiple package sizes up to 500 g to support scale-up.
  • Commonly used as a building block and intermediate in organic synthesis.
Catalog No. 50-003-02123 Supplier Medchemexpress LLC Supplier No. HYW014789500G
Diethyl propylmalonate (CAS 2163-48-6) is an organic diester used as a synthetic intermediate in medicinal chemistry and in pesticide synthesis. It is supplied as a high-purity reagent for research, development, and organic synthesis workflows.
  • High purity (99.7%) suitable for synthetic applications.
  • Molecular formula C10H18O4 and molecular weight 202.25 g/mol.
  • Offered in multiple package sizes up to 500 g to support scale-up.
  • Commonly used as a building block and intermediate in organic synthesis.