Learn More
Medchemexpress LLC Diethyl pyridine-2,4-dicarboxylate | 41438-38-4 | 99.8% | 223.23 g/mol | C11H13NO4 | 50 MG
SDP
Catalog No. 5000301620
Encompass_Preferred
Diethyl pyridine-2,4-dicarboxylate is a small-molecule proinhibitor directed at prolyl 4-hydroxylase that inhibits prolyl hydroxylation and procollagen processing in chick-embryo calvaria. It is supplied as a high-purity solid for research use only.
  • Prolyl 4-hydroxylase-directed proinhibitor with demonstrated biochemical activity.
  • High purity (99.8%) suitable for biochemical and developmental studies.
  • Solid form convenient for storage and compound handling.
  • Available in small pack sizes for screening and assay development.
  • Supplied with SDS for safe handling and storage guidance.
  • For research use only; not intended for human therapeutic use.
Catalog No. 50-003-01620 Supplier Medchemexpress LLC Supplier No. HYW02506250MG
Diethyl pyridine-2,4-dicarboxylate is a small-molecule proinhibitor directed at prolyl 4-hydroxylase that inhibits prolyl hydroxylation and procollagen processing in chick-embryo calvaria. It is supplied as a high-purity solid for research use only.
  • Prolyl 4-hydroxylase-directed proinhibitor with demonstrated biochemical activity.
  • High purity (99.8%) suitable for biochemical and developmental studies.
  • Solid form convenient for storage and compound handling.
  • Available in small pack sizes for screening and assay development.
  • Supplied with SDS for safe handling and storage guidance.
  • For research use only; not intended for human therapeutic use.