Learn More
Medchemexpress LLC Diethyl pyridine-2,4-dicarboxylate | 41438-38-4 | MFCD00152168 | 99.8% | 223.23 g/mol | C11H13NO4 | 1 ML
SDP
Catalog No. 5000301609
Encompass_Preferred
Diethyl pyridine-2,4-dicarboxylate is a small-molecule prolyl 4-hydroxylase-directed proinhibitor used to inhibit prolyl hydroxylation and procollagen processing in biological studies. It is offered as both a solid and as a ready-to-use 10 mM solution in DMSO for screening and cellular assays. Molecular formula C11H13NO4; molecular weight 223.23 g/mol.
Catalog No. 50-003-01609 Supplier Medchemexpress LLC Supplier No. HYW02506210MM/1ML
Diethyl pyridine-2,4-dicarboxylate is a small-molecule prolyl 4-hydroxylase-directed proinhibitor used to inhibit prolyl hydroxylation and procollagen processing in biological studies. It is offered as both a solid and as a ready-to-use 10 mM solution in DMSO for screening and cellular assays. Molecular formula C11H13NO4; molecular weight 223.23 g/mol.