Learn More
Medchemexpress LLC Diethyl pyridine-2,4-dicarboxylate | 41438-38-4 | MFCD00152168 | 99.8% | 223.23 g/mol | C11H13NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW025062100MG
Encompass_Preferred
Diethyl pyridine-2,4-dicarboxylate (CAS 41438-38-4) is a small-molecule proinhibitor of prolyl 4-hydroxylase used in biochemical research to inhibit prolyl hydroxylation and procollagen processing. It is supplied as an off-white to light-yellow solid with high purity suitable for laboratory studies.
  • Inhibits prolyl hydroxylation and procollagen processing in biological assays.
  • High purity for reproducible experimental results.
  • Solid form allows flexible formulation and storage.
  • Suitable for biochemical and cellular research applications.
  • Characterized by a defined molecular formula and molecular weight for identification.
Catalog No. 50-003-00455