Learn More
Crescent Chemical Co Inc DIETHYL PYROCARBONATE
SDP
Catalog No. 50247500
Encompass_Preferred
Diethyl pyrocarbonate research grade Crosslinks proteins, reacts with histidine residues (1). RNase inhibitor (2). Sterilization of instruments: 20-minute treatment with a dilution of 1 ml/l (3). Removal of RNases from solutions (except those containing amines like Tri
Catalog No. 50-247-500 Supplier Crescent Chemical Co Inc Supplier No. SE18835.01
Diethyl pyrocarbonate research grade Crosslinks proteins, reacts with histidine residues (1). RNase inhibitor (2). Sterilization of instruments: 20-minute treatment with a dilution of 1 ml/l (3). Removal of RNases from solutions (except those containing amines like Tri