Learn More
Medchemexpress LLC Diethyl pyrocarbonate | 1609-47-8 | 98.0% | 500 G
SDP
Catalog No. 5000410378
Encompass_Preferred
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It is useful in vitro as an agent that binds relatively specifically to the imidazole of histidine. Diethyl pyrocarbonate inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be utilized for modeling purposes.
  • Potent chemical inhibitor of RNase
  • Useful for in vitro applications
  • Binds specifically to imidazole of histidine
  • Inhibits central chemosensitivity
  • Modifies Ser, Thr, His, and Tyr residues
Catalog No. 50-004-10378 Supplier Medchemexpress LLC Supplier No. HYD0846500G
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It is useful in vitro as an agent that binds relatively specifically to the imidazole of histidine. Diethyl pyrocarbonate inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be utilized for modeling purposes.
  • Potent chemical inhibitor of RNase
  • Useful for in vitro applications
  • Binds specifically to imidazole of histidine
  • Inhibits central chemosensitivity
  • Modifies Ser, Thr, His, and Tyr residues