Learn More
Medchemexpress LLC Diethyl pyrocarbonate | 1609-47-8 | 99.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD084610MM/1ML
Encompass_Preferred
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. Diethyl pyrocarbonate inhibits central chemosensitivity in rabbits. It can modify Ser, Thr, His, and Tyr residues, and can be used for modeling.
  • Potent, orally active, non-specific chemical inhibitor of RNase.
  • Useful in vitro for binding to imidazole of histidine.
  • Inhibits central chemosensitivity in rabbits.
  • Capable of modifying Ser, Thr, His, and Tyr residues.
  • Can be used for modeling.
Catalog No. 50-004-09839