Learn More
Medchemexpress LLC Diethyl pyrocarbonate | 1609-47-8 | 99.3% | 100 G
SDP
Catalog No. 5000409973
Encompass_Preferred
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. It inhibits central chemosensitivity in rabbits. It can modify Ser, Thr, His, and Tyr residues and be used for modeling. This product is for research use only.
  • Potent, orally active, non-specific chemical inhibitor of RNase.
  • Useful in vitro as an agent relatively specific for binding to imidazole of histidine.
  • Inhibits central chemosensitivity in rabbits.
  • Can modify Ser, Thr, His, and Tyr residues.
  • Can be used for modeling.
Catalog No. 50-004-09973 Supplier Medchemexpress LLC Supplier No. HYD0846100G
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. It inhibits central chemosensitivity in rabbits. It can modify Ser, Thr, His, and Tyr residues and be used for modeling. This product is for research use only.
  • Potent, orally active, non-specific chemical inhibitor of RNase.
  • Useful in vitro as an agent relatively specific for binding to imidazole of histidine.
  • Inhibits central chemosensitivity in rabbits.
  • Can modify Ser, Thr, His, and Tyr residues.
  • Can be used for modeling.