Learn More
Medchemexpress LLC Diethyl pyrocarbonate | 1609-47-8 | 99.3% | 50 G
SDP
Catalog No. 5000409959
Encompass_Preferred
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. Diethyl pyrocarbonate also inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be utilized for modeling purposes.
  • Potent and orally active.
  • Non-specific chemical inhibitor of RNase.
  • Relatively specific for binding to imidazole of histidine in vitro.
  • Inhibits central chemosensitivity.
  • Modifies Ser, Thr, His, and Tyr residues.
  • Can be used for modeling.
Catalog No. 50-004-09959 Supplier Medchemexpress LLC Supplier No. HYD084650G
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. Diethyl pyrocarbonate also inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be utilized for modeling purposes.
  • Potent and orally active.
  • Non-specific chemical inhibitor of RNase.
  • Relatively specific for binding to imidazole of histidine in vitro.
  • Inhibits central chemosensitivity.
  • Modifies Ser, Thr, His, and Tyr residues.
  • Can be used for modeling.