Learn More
Medchemexpress LLC Diethyl pyrocarbonate | 1609-47-8 | 99.33% | 250 G
SDP
Catalog No. 5000414483
Encompass_Preferred
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. It inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be used for modeling.
  • Potent, orally active, non-specific chemical inhibitor of RNase.
  • Useful in vitro as an agent relatively specific for binding to imidazole of histidine.
  • Inhibits central chemosensitivity in rabbits.
  • Can modify Ser, Thr, His and Tyr residues.
  • Can be used for modeling.
Catalog No. 50-004-14483 Supplier Medchemexpress LLC Supplier No. 250GHYD0846
Diethyl pyrocarbonate is a potent, orally active, non-specific chemical inhibitor of RNase. It has been useful in vitro as an agent relatively specific for binding to imidazole of histidine. It inhibits central chemosensitivity in rabbits and can modify Ser, Thr, His, and Tyr residues. It can be used for modeling.
  • Potent, orally active, non-specific chemical inhibitor of RNase.
  • Useful in vitro as an agent relatively specific for binding to imidazole of histidine.
  • Inhibits central chemosensitivity in rabbits.
  • Can modify Ser, Thr, His and Tyr residues.
  • Can be used for modeling.