Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl pyrocarbonate, 97%, pure, ACROS Organics™

Catalog No. AC170250025
Change view
Click to view available options
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC170250025 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170250025 Supplier Thermo Scientific Chemicals Supplier No. 170250025

missing translation for 'validMainframeMsg'

Specifications

Specific Gravity 1.1175
Viscosity 1.97 mPa.s (20°C)
Flash Point 69°C
Quantity 2.5kg
Refractive Index 1.3960 to 1.4000
Density 1.1175g/mL
Packaging Glass bottle
Boiling Point 93.0°C to 94.0°C (18.0mmHg)
CAS 1609-47-8
Assay Percent Range 0.97
Molecular Formula C6H10O5
Linear Formula O(CO2C2H5)2
MDL Number MFCD00009106
Beilstein 03,IV,18
Merck Index 153146
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
Solubility Information Solubility in water: slow decomposition. Other solubilities: very soluble in acetone,ethanol,ligroin
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILES CCOC(=O)OC(=O)OCC
IUPAC Name ethoxycarbonyl ethyl carbonate
Molecular Weight (g/mol) 162.14
PubChem CID 3051
ChEBI CHEBI:59051
Formula Weight 162.14
Percent Purity 97%
Grade Pure
Chemical Name or Material Diethyl pyrocarbonate
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Flammable liquids (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Harmful if swallowed, Combustible liquid, May cause respiratory irritation, Causes skin irritation
EINECSNumber 216-542-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.