Learn More
Medchemexpress LLC Diethyl-pythiDC | 1821370-70-0 | 99.90% | 306.34 | 10 MG
SDP
Catalog No. 5000364836
Encompass_Preferred
Diethyl-pythiDC is an inhibitor of collagen prolyl 4-hydroxylases (CP4Hs). This product is intended for laboratory chemical use and for the manufacture of substances.
  • Inhibitor of collagen prolyl 4-hydroxylases (CP4Hs).
  • For research use only.
  • Does not cause iron deficiency in cultured cells.
  • No cytotoxic activity at high micromolar concentrations in MDA-MB-231 cells.
  • Soluble in DMSO (33.33 mg/mL).
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Solution storage: -80°C for 2 years; -20°C for 1 year.
  • Appears as an off-white to light yellow solid.
Catalog No. 50-003-64836 Supplier Medchemexpress LLC Supplier No. 10MGHY103068
Diethyl-pythiDC is an inhibitor of collagen prolyl 4-hydroxylases (CP4Hs). This product is intended for laboratory chemical use and for the manufacture of substances.
  • Inhibitor of collagen prolyl 4-hydroxylases (CP4Hs).
  • For research use only.
  • Does not cause iron deficiency in cultured cells.
  • No cytotoxic activity at high micromolar concentrations in MDA-MB-231 cells.
  • Soluble in DMSO (33.33 mg/mL).
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Solution storage: -80°C for 2 years; -20°C for 1 year.
  • Appears as an off-white to light yellow solid.