Learn More
Medchemexpress LLC Diethyl-pythiDC | 1821370-70-0 | MFCD31715412 | 99.9% | 306.34 g/mol | C14H14N2O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10306825MG
Encompass_Preferred
Diethyl-pythiDC is a small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs), active in cell-based assays at concentrations that do not induce iron deficiency. It is provided for research use as a high-purity solid and in DMSO solution formats.
  • Inhibits collagen prolyl 4-hydroxylases (CP4Hs) in cultured cells.
  • Active at concentrations that do not cause iron deficiency.
  • Provided as a high-purity research reagent (99.90%).
  • Available in smaller research pack sizes and as a DMSO solution for assay use.
Catalog No. 50-002-46147