Learn More
Medchemexpress LLC Diethyl succinate | 123-25-1 | MFCD00009208 | 98.0% | 174.19 g/mol | C8H14O4 | 1mL
SDP
Catalog No. 5000268625
Encompass_Preferred
Diethyl succinate (Diethyl Butanedioate) can be utilized at physiological pH allowing it to penetrate biological membranes and integrate into the cells of tissue cultures where it is metabolized via the tricarboxylic acid cycle Diethyl succinate modulates the polarization and activation of microglial cells by reducing mitochondrial fission and the levels of reactive oxygen species (ROS) thereby exerting an inflammatory protective effect in primary microglial cells Furthermore Diethyl succinate is non-toxic and can be used in flavorings and seasonings[1][2]
Catalog No. 50-002-68625 Supplier Medchemexpress LLC Supplier No. HYY083610MM/1ML
Diethyl succinate (Diethyl Butanedioate) can be utilized at physiological pH allowing it to penetrate biological membranes and integrate into the cells of tissue cultures where it is metabolized via the tricarboxylic acid cycle Diethyl succinate modulates the polarization and activation of microglial cells by reducing mitochondrial fission and the levels of reactive oxygen species (ROS) thereby exerting an inflammatory protective effect in primary microglial cells Furthermore Diethyl succinate is non-toxic and can be used in flavorings and seasonings[1][2]