Learn More
Medchemexpress LLC Diethyl succinate-13C4 | 1628796-56-4 | MFCD00009208 | 178.16 | C8H14O4 (1,2,3,4-13C4) | 5mg
SDP
Catalog No. 5000268626
Encompass_Preferred
Diethyl succinate-13C4 is the 13C labeled Diethyl succinate[1] Diethyl succinate (Diethyl Butanedioate) is used at physiological pH and crosses biological membranes incorporates into cells in tissue culture and is metabolized by the TCA cycle Diethyl succinate is known to be non-toxic and used in fragrances and flavoring[2]
Catalog No. 50-002-68626 Supplier Medchemexpress LLC Supplier No. HYY0836S5MG
Diethyl succinate-13C4 is the 13C labeled Diethyl succinate[1] Diethyl succinate (Diethyl Butanedioate) is used at physiological pH and crosses biological membranes incorporates into cells in tissue culture and is metabolized by the TCA cycle Diethyl succinate is known to be non-toxic and used in fragrances and flavoring[2]