Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl trans-1,2-Cyclohexanedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D36845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36845G Supplier TCI America Supplier No. D36845G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17351-22-3
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.288
InChI Key ZTUZDYWYNQDJKR-NXEZZACHSA-N
Synonym trans-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester
PubChem CID 44629905
IUPAC Name diethyl (1R,2R)-cyclohexane-1,2-dicarboxylate
SMILES CCOC(=O)C1CCCCC1C(=O)OCC

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl trans-1,2-Cyclohexanedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.