Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl trans-4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™
SDP

Catalog No. C17041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17041G 1 g
C17045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17041G Supplier TCI America Supplier No. C17041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5048-50-0
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD06799457
InChI Key OWVOHDKOANTMJU-UHFFFAOYNA-N
Synonym trans-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester, Diethyl trans-1,2,3,6-Tetrahydrophthalate, trans-1,2,3,6-Tetrahydrophthalic Acid Diethyl Ester
PubChem CID 12358152
IUPAC Name 1,2-diethyl cyclohex-4-ene-1,2-dicarboxylate
SMILES CCOC(=O)C1CC=CCC1C(=O)OCC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 226.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl trans-4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.