Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylaluminum Chloride Solution

Catalog No. 212806800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-280-6800ML 800 mL
21-280-625ML 4 x 25 mL
21-280-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 21-280-6800ML Supplier Sigma Aldrich Supplier No. 212806800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylaluminum chloride solution can be used:As a Lewis acid for the synthesis of Lewis acid-base bifunctional asymmetric catalysts for cyanosilylation of aldehydes and in Strecker reaction.For the synthesis of precursors for lanthanide/aluminum catalysts for diene polymerization.As a co-initiator in the cationic polymerization of isobutylene.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 96-10-6
Density 0.711 g/mL at 25°C
Molecular Formula C4H10AlCl
Linear Formula (C2H5)2AlCl
MDL Number MFCD00000459
Quantity 800 mL
Molecular Weight (g/mol) 120.56 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.