Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylaluminum Cyanide Solution

Catalog No. 276863800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-686-3800ML 800 mL
27-686-3100ML 100 mL
50-308-1493 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 27-686-3800ML Supplier Sigma Aldrich Supplier No. 276863800ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used to hydrocyanate side chain ketones directed by aryl sulfinyl groups.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 5804-85-3
Density 0.864 g/mL at 25°C
Molecular Formula C5H10AlN
Linear Formula (C2H5)2AlCN
MDL Number MFCD00001764
Quantity 800 mL
Molecular Weight (g/mol) 111.12 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.