Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Diethylamine hydrochloride | 2KG | 660-68-4 | MFCD00012499

Catalog No. 12774425KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

CAS #: 660-68-4
MDL #: MFCD00012499
Purity: >99 %
UNSPSC Code: 12352100
Molecular Weight: 109.6

Catalog No. 12-774-425KG Supplier Sigma Organic Chemistry Supplier No. 1277442.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #: 660-68-4
MDL #: MFCD00012499
Purity: >99 %
UNSPSC Code: 12352100
Molecular Weight: 109.6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.