Learn More
Medchemexpress LLC Diethylamine nonoate (diethylammonium salt) | 372965-00-9 | 98.0% | C8H22N4O2 | 10MG
SDP
Catalog No. 5000261099
Encompass_Preferred
Diethylamine nonoate (diethylammonium salt) is a nitric oxide (NO) donor used in biochemical and pharmacological research. It is a crystalline solid (CAS 372965-00-9) with reported purity of 98.0% and molecular weight 206.29, typically stored at low temperatures to preserve activity.
  • Releases nitric oxide under physiological conditions.
  • High reported purity for reproducible experimental results.
  • Solid formulation suitable for accurate weighing and dosing.
  • Applicable to studies of vasodilation, cell signaling, and nitrosative stress.
  • Stable when stored at recommended low temperatures.
Catalog No. 50-002-61099 Supplier Medchemexpress LLC Supplier No. HY13192510MG
Diethylamine nonoate (diethylammonium salt) is a nitric oxide (NO) donor used in biochemical and pharmacological research. It is a crystalline solid (CAS 372965-00-9) with reported purity of 98.0% and molecular weight 206.29, typically stored at low temperatures to preserve activity.
  • Releases nitric oxide under physiological conditions.
  • High reported purity for reproducible experimental results.
  • Solid formulation suitable for accurate weighing and dosing.
  • Applicable to studies of vasodilation, cell signaling, and nitrosative stress.
  • Stable when stored at recommended low temperatures.