Learn More
Medchemexpress LLC HY-N6658 100mg Medchemexpress, 6-Hydroxy-4-methylcoumarin CAS:2373-31-1 Purity:>98%
SDP
Catalog No. 501970452
Encompass_Preferred
Medchemexpress, HY-109656 100mg Diethylamino hydroxybenzoyl hexyl benzoate CAS:302776-68-7 Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0452 Supplier Medchemexpress LLC Supplier No. HYN6658100MG
Medchemexpress, HY-109656 100mg Diethylamino hydroxybenzoyl hexyl benzoate CAS:302776-68-7 Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.